missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBX4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18544-1-AP-150UL
This item is not returnable.
View return policy
Description
Polycomb group (PcG) proteins are chromatin-associated proteins which are important for embryonic and adult stem cell self-renewal and maintece. At least two distinct human PcG complexes have been identified. Polycomb repressive complex I (PRC1) contains CBX4/Pc2, HPH1 and RING domain-containing proteins (RING1, RING2 and BMI1). CBX4 (Chromobox protein homolog 4 or PC2), which functions as a transcriptional suppressor, is a candidate of KyoT2-binding proteins. It is a E3 SUMO-protein ligase which facilitates SUMO1 conjugation. Depletion of CBX4 results in decreased cellular resistance to ionizing radiation. CBX4 is directly involved in the cellular response to DNA damage.Specifications
| CBX4 | |
| Polyclonal | |
| Unconjugated | |
| CBX4 | |
| CBX4, Chromobox protein homolog 4, E3 SUMO protein ligase CBX4, hPC2, NBP16, PC2, Polycomb 2 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12418, 501403, 8535 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00257, O55187 | |
| Cbx4 | |
| CBX4 Fusion Protein Ag13425 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction