missing translation for 'onlineSavingsMsg'
Learn More

CCK-AR Antibody (377251), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18526787 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18526787 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18526787 Supplier Novus Biologicals Supplier No. FAB2680J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCK-AR Monoclonal antibody specifically detects CCK-AR in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCK-AR
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 377251
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CCK1-R, CCK-A, CCK-A receptor, CCK-AR, CCKRA, cholecystokinin A receptor, cholecystokinin receptor type A, cholecystokinin type-A receptor, Cholecystokinin-1 receptor
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CCK-A R, Met1-Gln428, Accession # P32238
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, GPCR, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 886
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.