missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR3 Antibody (8E9H10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61672-0.025ml
This item is not returnable.
View return policy
Description
CCR3 Monoclonal specifically detects CCR3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| CCR3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR-3, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841 | |
| Mouse | |
| 41 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA | |
| 8E9H10 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CCR3 | |
| Purified recombinant fragment of human CD193 (AA: extra mix) expressed in E. Coli. | |
| Protein G purified | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1232 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction