missing translation for 'onlineSavingsMsg'
Learn More

CCR3 Antibody (8E9H10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

203.00€ - 557.00€

Specifications

Antigen CCR3
Clone 8E9H10
Concentration 1 mg/mL
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Applications Western Blot, Flow Cytometry, ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18223534
View Documents
Novus Biologicals
NBP2-61672
0.1 mL
557.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18615427
View Documents
Novus Biologicals
NBP2-61672-0.025ml
0.025 mL
203.00€
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CCR3 Monoclonal specifically detects CCR3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

CCR3
1 mg/mL
Western Blot, Flow Cytometry, ELISA
Unconjugated
Mouse
b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR-3, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841
CCR3
IgG1
Protein G purified
8E9H10
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Monoclonal
Purified
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
1232
Purified recombinant fragment of human CD193 (AA: extra mix) expressed in E. Coli.
Primary
41 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.