missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCRL2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13387-1-AP-150UL
This item is not returnable.
View return policy
Description
CCRL2 (C-C chemokine receptor-like 2), also known as chemokine receptor X, human chemokine receptor (HCR), CRAM-A or CRAM-B, is a seven pass transmembrane protein expressed by monocytes, neutrophils, and dendritic cells of immunal tissues (predomily spleen, fetal liver, bone marrow and lymph node). It shares over 40% homology with other C-C chemokine receptors but does not share the conserved DRY motif which is among the important motifs necessary for signalling and ligand-binding. C-C chemokine receptors are G-protein coupled, seven pass transmembrane domain proteins whose major physiological role is to function in the chemotaxis of T cells and phagocytic cells to areas of inflammation.Specifications
| CCRL2 | |
| Polyclonal | |
| Unconjugated | |
| CCRL2 | |
| C C chemokine receptor like 2, CCR11, CCR6, CCRL2, Chemokine receptor CCR11, Chemokine receptor X, CKRX, CRAM, CRAM A, CRAM B, HCR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54199, 9034 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00421, O35457 | |
| Ccrl2 | |
| CCRL2 Fusion Protein Ag4045 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction