missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCRL2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | CCRL2 |
|---|---|
| Concentration | 0.21 mg/mL |
| Applications | Western Blot, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16821574
|
Proteintech
13387-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16811574
|
Proteintech
13387-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCRL2 (C-C chemokine receptor-like 2), also known as chemokine receptor X, human chemokine receptor (HCR), CRAM-A or CRAM-B, is a seven pass transmembrane protein expressed by monocytes, neutrophils, and dendritic cells of immunal tissues (predomily spleen, fetal liver, bone marrow and lymph node). It shares over 40% homology with other C-C chemokine receptors but does not share the conserved DRY motif which is among the important motifs necessary for signalling and ligand-binding. C-C chemokine receptors are G-protein coupled, seven pass transmembrane domain proteins whose major physiological role is to function in the chemotaxis of T cells and phagocytic cells to areas of inflammation.Specifications
| CCRL2 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| O00421, O35457 | |
| 54199, 9034 | |
| CCRL2 Fusion Protein Ag4045 | |
| Primary | |
| -20°C | |
| CCRL2 |
| 0.21 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| C C chemokine receptor like 2, CCR11, CCR6, CCRL2, Chemokine receptor CCR11, Chemokine receptor X, CKRX, CRAM, CRAM A, CRAM B, HCR | |
| Ccrl2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title