missing translation for 'onlineSavingsMsg'
Learn More

CD117 / c-Kit Rat, APC-Cy7, Clone: ACK2, enQuireBio™

Rat Monoclonal Antibody

248.85€ - 433.65€

Specifications

Antigen CD117 / c-Kit
Clone ACK2
Concentration 0.2mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15922188
View Documents
EnquireBio
QAB54-APC7-25ug
25 μg
248.85€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15912188
View Documents
EnquireBio
QAB54-APC7-100ug
100 μg
433.65€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD117 / c-Kit
0.2mg/mL
Monoclonal
Rat
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
16590
IgG2b κ
Protein A/G Purified
ACK2
Flow Cytometry
APC-Cyanine7
RUO
NP_066922
Kit
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.