missing translation for 'onlineSavingsMsg'
Learn More

EnquireBio CD117 / c-Kit Rat, APC-Cy7, Clone: ACK2, enQuireBio™

Rat Monoclonal Antibody

Brand:  EnquireBio QAB54-APC7-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15922188

  • 188.00€ / 25µg

Estimated Shipment: 02-08-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

CD117 / c-Kit
Monoclonal
0.2mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Kit
Protein A/G Purified
RUO
16590
IgG2b κ
Flow Cytometry
ACK2
APC-Cyanine7
NP_066922
Rat
25 μg
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions