Learn More
CD150 (SLAM) Hamster, anti-Mouse, PE-Cyanine7, Clone: TC15-12F12.2, STEMCELL Technologies
Hamster Monoclonal Antibody
Brand: Stemcell Technologies 60036CZ
Description
- The TC15-12F12
- 2 antibody reacts with CD150 (signaling lymphocyte activation molecule or SLAM), an ∼75 kDa type I transmembrane glycoprotein that plays multiple roles in the immune response by serving as a cell adhesion molecule and/or coreceptor
- It is differentially expressed by T cells, immature thymocytes, B cells, dendritic cells, macrophages, and endothelial cells
- The expression pattern differs according to cell type and activation status
- Expression is rapidly induced upon activation of T cells, B cells and dendritic cells, with synthesis by T cells being maintained on Th1 but not Th2 clones
- CD150-mediated co-stimulation of TCR-activated T cells enhances the production of IFN-γ by Th1 cells, a response that can be augmented by binding of the TC15-12F12
- 2 antibody
- CD150 is thought to mediate signal transduction by associating with the intracellular protein tyrosine phosphatase, SHP-2
Specifications
| CD150 (SLAM) | |
| Monoclonal | |
| PE-Cyanine7 | |
| 100 μg | |
| Mouse | |
| Hematopoietic Stem and Progenitor Cells |
| Immunohistochemistry, Flow Cytometry, Immunofluorescence, Functional Assay, Immunoprecipitation | |
| TC15-12F12.2 | |
| Hamster | |
| Primary | |
| IgG2a λ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.