Learn More
CD150 (SLAM) Hamster, anti-Mouse, PE-Cyanine7, Clone: TC15-12F12.2, STEMCELL Technologies
Hamster Monoclonal Antibody
Brand:  Stemcell Technologies 60036CZ
Encompass_Preferred
 View more versions of this product
Product Code. 17178681
  • 785.70€ / 100µg
Description
  • The TC15-12F12
  • 2 antibody reacts with CD150 (signaling lymphocyte activation molecule or SLAM), an ∼75 kDa type I transmembrane glycoprotein that plays multiple roles in the immune response by serving as a cell adhesion molecule and/or coreceptor
  • It is differentially expressed by T cells, immature thymocytes, B cells, dendritic cells, macrophages, and endothelial cells
  • The expression pattern differs according to cell type and activation status
  • Expression is rapidly induced upon activation of T cells, B cells and dendritic cells, with synthesis by T cells being maintained on Th1 but not Th2 clones
  • CD150-mediated co-stimulation of TCR-activated T cells enhances the production of IFN-γ by Th1 cells, a response that can be augmented by binding of the TC15-12F12
  • 2 antibody
  • CD150 is thought to mediate signal transduction by associating with the intracellular protein tyrosine phosphatase, SHP-2
Specifications
Specifications
CD150 (SLAM)
Monoclonal
PE-Cyanine7
100 μg
Mouse
Hematopoietic Stem and Progenitor Cells
Immunohistochemistry, Flow Cytometry, Immunofluorescence, Functional Assay, Immunoprecipitation
TC15-12F12.2
Hamster
Primary
IgG2a λ