missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD150 (SLAM) Hamster, anti-Mouse, PE-Cyanine7, Clone: TC15-12F12.2, STEMCELL Technologies
Hamster Monoclonal Antibody
284.85€ - 785.70€
Specifications
| Antigen | CD150 (SLAM) |
|---|---|
| Clone | TC15-12F12.2 |
| Applications | Immunohistochemistry, Flow Cytometry, Immunofluorescence, Functional Assay, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | PE-Cyanine7 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17188681
|
Stemcell Technologies
60036CZ.1 |
25 μg |
284.85€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17178681
|
Stemcell Technologies
60036CZ |
100 μg |
785.70€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The TC15-12F12
- 2 antibody reacts with CD150 (signaling lymphocyte activation molecule or SLAM), an ∼75 kDa type I transmembrane glycoprotein that plays multiple roles in the immune response by serving as a cell adhesion molecule and/or coreceptor
- It is differentially expressed by T cells, immature thymocytes, B cells, dendritic cells, macrophages, and endothelial cells
- The expression pattern differs according to cell type and activation status
- Expression is rapidly induced upon activation of T cells, B cells and dendritic cells, with synthesis by T cells being maintained on Th1 but not Th2 clones
- CD150-mediated co-stimulation of TCR-activated T cells enhances the production of IFN-γ by Th1 cells, a response that can be augmented by binding of the TC15-12F12
- 2 antibody
- CD150 is thought to mediate signal transduction by associating with the intracellular protein tyrosine phosphatase, SHP-2
Specifications
| CD150 (SLAM) | |
| Immunohistochemistry, Flow Cytometry, Immunofluorescence, Functional Assay, Immunoprecipitation | |
| PE-Cyanine7 | |
| Mouse | |
| Primary |
| TC15-12F12.2 | |
| Monoclonal | |
| Hamster | |
| IgG2a λ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title