missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD16 Mouse, anti-Human, FITC, Clone: 3G8, STEMCELL Technologies
Mouse Monoclonal Antibody
193.05€ - 433.35€
Specifications
| Antigen | CD16 |
|---|---|
| Clone | 3G8 |
| Applications | Immunohistochemistry, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17108741
|
Stemcell Technologies
60041FI.1 |
25 Tests |
193.05€
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17198731
|
Stemcell Technologies
60041FI |
100 Tests |
433.35€
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The 3G8 antibody reacts with CD16 (FcγRIII or low affinity IgG receptor III), a type 1 transmembrane glycoprotein belonging to the Ig superfamily
- CD16 exists in two distinct isoforms: CD16a, a 50 - 65 kDa form expressed on NK cells, activated monocytes, macrophages, and placental trophoblasts, and CD16b, an ∼48 kDa glycosylphosphatidylinositol (GPI)-anchored form expressed on neutrophils, basophils and eosinophils and found as at least two polymorphic variants, termed NA1 and NA2
- CD16 binds weakly to the Fc region of monomeric, aggregated or complexed IgG, particularly the IgG1 and IgG3 isotypes
- Binding of IgG to either CD16 isoform induces signaling pathways that modulate several types of responses, including antibody-dependent cell-mediated cytotoxicity (ADCC), phagocytosis, cytokine release and proliferation
- CD16/IgG interactions can result in non-specific labeling in antibody-based detection and cell separation experiments
Specifications
| CD16 | |
| Immunohistochemistry, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| FITC | |
| Baboon, Cynomolgus, Chimpanzee, Human | |
| Primary |
| 3G8 | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title