Learn More
CD25 Rat, anti-Mouse, PE, Clone: PC61.5, STEMCELL Technologies
Rat Monoclonal Antibody
Brand:  Stemcell Technologies 60009PE
Encompass_Preferred
 View more versions of this product
Product Code. 17168401
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • The PC61.5 antibody reacts with mouse CD25 (low-affinity interleukin‐2 receptor α chain or IL-2Rα), which is an ∼55 kDa type 1 transmembrane glycoprotein expressed on T and B cell progenitors, activated (but not resting) T and B cells, regulatory T (Treg) cells, dendritic cells, and activated monocytes and macrophages
  • Expression of CD25, together with CD4 and FOXP3, is considered a phenotypic signature for Treg cells
  • CD25 per se has low affinity for its IL-2 ligand but associates with CD122 (IL-2Rβ) and CD132 (IL-2Rγ) to form the high-affinity IL-2R receptor
  • CD25 is necessary for receptor clustering and induction of signaling pathways involved in the activation and proliferation of lymphocytes
  • Binding of the PC61.5 antibody inhibits binding of IL-2 to the low- and high-affinity receptors, which may result in inhibition of IL-2-dependent proliferation
  • The epitope recognized by PC61.5 is distinct from the IL-2 binding site and from the epitopes recognized by antibodies 7D4 and 3C7
Specifications
Specifications
CD25
Monoclonal
PE
100 μg
Mouse
T Cells, T Cells, Regulatory
Immunohistochemistry, Flow Cytometry, Functional Assay, Blocking Assay
PC61.5
Rat
Primary
IgG1 λ