missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25 Rat, anti-Mouse, PE, Clone: PC61.5, STEMCELL Technologies
Rat Monoclonal Antibody
POA - 110.70€
Specifications
| Antigen | CD25 |
|---|---|
| Clone | PC61.5 |
| Applications | Immunohistochemistry, Flow Cytometry, Functional Assay, Blocking Assay |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17178401
|
Stemcell Technologies
60009PE.1 |
25 μg |
110.70€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17168401
|
Stemcell Technologies
60009PE |
100 μg | |||||||
Description
- The PC61.5 antibody reacts with mouse CD25 (low-affinity interleukin‐2 receptor α chain or IL-2Rα), which is an ∼55 kDa type 1 transmembrane glycoprotein expressed on T and B cell progenitors, activated (but not resting) T and B cells, regulatory T (Treg) cells, dendritic cells, and activated monocytes and macrophages
- Expression of CD25, together with CD4 and FOXP3, is considered a phenotypic signature for Treg cells
- CD25 per se has low affinity for its IL-2 ligand but associates with CD122 (IL-2Rβ) and CD132 (IL-2Rγ) to form the high-affinity IL-2R receptor
- CD25 is necessary for receptor clustering and induction of signaling pathways involved in the activation and proliferation of lymphocytes
- Binding of the PC61.5 antibody inhibits binding of IL-2 to the low- and high-affinity receptors, which may result in inhibition of IL-2-dependent proliferation
- The epitope recognized by PC61.5 is distinct from the IL-2 binding site and from the epitopes recognized by antibodies 7D4 and 3C7
Specifications
| CD25 | |
| Immunohistochemistry, Flow Cytometry, Functional Assay, Blocking Assay | |
| PE | |
| Mouse | |
| Primary |
| PC61.5 | |
| Monoclonal | |
| Rat | |
| IgG1 λ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title