missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD31/PECAM-1 Antibody (CO.3E-1D4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB100-65900-0.025MG
This item is not returnable.
View return policy
Description
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Bovine, Equine, Goat, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| CD31/PECAM-1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1:10-1:25, Immunohistochemistry 1:50, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:50 | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet/endothelial cell adhesion molecule | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-65900 is specific for ovine CD31, also known as PECAM-1. Ovine CD31 is predominantly expressed by peripheral blood platelets and a small percentage of lymphocytes. CD31 is also highly expressed by ovine endothelial cells. Clone CO.3E1D4 is reported to inhibit homotypic leucocyte aggregation induced by anti CD43 antibodies (2). Removal of sodium azide is recommended prior to use in fuctional assays. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Immunohistochemistry (Frozen), Immunohistochemistry, Flow Cytometry, Immunoprecipitation | |
| CO.3E-1D4 | |
| Unconjugated | |
| Phosphate buffered saline with 0.09% Sodium Azide | |
| PECAM1 | |
| Ovine leucocytes | |
| 0.025 mg | |
| Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 5175 | |
| Goat, Sheep, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction