missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD31/PECAM-1 Antibody (CO.3E-1D4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
222.00€ - 537.00€
Specifications
| Antigen | CD31/PECAM-1 |
|---|---|
| Clone | CO.3E-1D4 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:10-1:25, Immunohistochemistry 1:50, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:50 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry, Flow Cytometry, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18232244
|
Novus Biologicals
NB100-65900-0.025MG |
0.025 mg |
222.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18709573
|
Novus Biologicals
NB100-65900-0.1mg |
0.1 mg |
537.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Bovine, Equine, Goat, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| CD31/PECAM-1 | |
| 1.0 mg/mL | |
| Immunohistochemistry (Frozen), Immunohistochemistry, Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| Phosphate buffered saline with 0.09% Sodium Azide | |
| 5175 | |
| Ovine leucocytes | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CO.3E-1D4 | |
| Flow Cytometry 1:10-1:25, Immunohistochemistry 1:50, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Goat, Sheep, Bovine | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet/endothelial cell adhesion molecule | |
| PECAM1 | |
| IgG2a | |
| Protein G purified | |
| NB100-65900 is specific for ovine CD31, also known as PECAM-1. Ovine CD31 is predominantly expressed by peripheral blood platelets and a small percentage of lymphocytes. CD31 is also highly expressed by ovine endothelial cells. Clone CO.3E1D4 is reported to inhibit homotypic leucocyte aggregation induced by anti CD43 antibodies (2). Removal of sodium azide is recommended prior to use in fuctional assays. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title