Learn More
CD4 Antibody (CE9.1 (Clenoliximab)) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52681-0.025MG
 View more versions of this product
Product Code. 18119169
  • 260.00€ / 25µg
Description
CD4 Monoclonal specifically detects CD4 in Human, Chimpanzee samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Surface Plasmon Resonance, CyTOF-ready, Block/Neutralize.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD4
Monoclonal
Unconjugated
PBS with 0.02% Proclin 300. with 0.02% Proclin 300
CD4
Domain 1 of human CD4.
0.025 mg
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Stem Cell Markers
920
Human, Primate
Purified
Flow Cytometry, ELISA, Immunohistochemistry, Blocking Assay, CyTOF
CE9.1 (Clenoliximab)
Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Rabbit
Protein A or G purified
RUO
Primary
The antibody binds to human CD4 with a Kd of 1.0 nM and an ED50 of 0.2 ug/ml.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.