missing translation for 'onlineSavingsMsg'
Learn More

CD4 Antibody (CE9.1 (Clenoliximab)) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody

260.00€ - 613.00€

Specifications

Antigen CD4
Clone CE9.1 (Clenoliximab)
Dilution Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready
Applications Flow Cytometry, ELISA, Immunohistochemistry, Blocking Assay, CyTOF
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18119169
View Documents
Novus Biologicals
NBP2-52681-0.025MG
0.025 mg
260.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18207950
View Documents
Novus Biologicals
NBP2-52681-0.2MG
0.2 mg
613.00€
0.20mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD4 Monoclonal specifically detects CD4 in Human, Chimpanzee samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Surface Plasmon Resonance, CyTOF-ready, Block/Neutralize.
Specifications

Specifications

CD4
Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready
Monoclonal
Purified
RUO
Human, Primate
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
CD4
IgG
Protein A or G purified
The antibody binds to human CD4 with a Kd of 1.0 nM and an ED50 of 0.2 ug/ml.
CE9.1 (Clenoliximab)
Flow Cytometry, ELISA, Immunohistochemistry, Blocking Assay, CyTOF
Unconjugated
Rabbit
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Stem Cell Markers
PBS with 0.02% Proclin 300. with 0.02% Proclin 300
920
Domain 1 of human CD4.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.