missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Antibody (CE9.1 (Clenoliximab)) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-52681-0.2MG
This item is not returnable.
View return policy
Description
CD4 Monoclonal specifically detects CD4 in Human, Chimpanzee samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Surface Plasmon Resonance, CyTOF-ready, Block/Neutralize.
Specifications
| CD4 | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| CD4 | |
| Domain 1 of human CD4. | |
| 0.2 mg | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Stem Cell Markers | |
| 920 | |
| Human, Primate | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Blocking Assay, CyTOF | |
| CE9.1 (Clenoliximab) | |
| Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready | |
| CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| The antibody binds to human CD4 with a Kd of 1.0 nM and an ED50 of 0.2 ug/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction