Learn More
CD4 Antibody (YNB46.1.8 (Campath-9H)) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52658-0.025MG
 View more versions of this product
Product Code. 18197689
  • 271.00€ / 25µg
Description
CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD4
Monoclonal
Unconjugated
PBS with 0.02% Proclin 300. with 0.02% Proclin 300
CD4
Rat T-cell line (NB2) transfected with the human CD4.
0.025 mg
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Stem Cell Markers
920
Human
Purified
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), CyTOF
YNB46.1.8 (Campath-9H)
Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, CyTOF-ready
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Rabbit
Protein A or G purified
RUO
Primary
Recognizes the 55kDa human CD4 cell surface antigen, expressed by helper T lymphocytes and by monocytes.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.