missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Antibody (YNB46.1.8 (Campath-9H)) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
271.00€ - 558.00€
Specifications
| Antigen | CD4 |
|---|---|
| Clone | YNB46.1.8 (Campath-9H) |
| Dilution | Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18197689
|
Novus Biologicals
NBP2-52658-0.025MG |
0.025 mg |
271.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18230960
|
Novus Biologicals
NBP2-52658-0.2MG |
0.2 mg |
558.00€
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, CyTOF-ready.Specifications
| CD4 | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| CD4 | |
| IgG | |
| Protein A or G purified | |
| Recognizes the 55kDa human CD4 cell surface antigen, expressed by helper T lymphocytes and by monocytes. |
| YNB46.1.8 (Campath-9H) | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), CyTOF | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Stem Cell Markers | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| 920 | |
| Rat T-cell line (NB2) transfected with the human CD4. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title