Learn More
CD4 Mouse, anti-Human, PerCP-Cyanine5.5, Clone: OKT4, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60016PS
Encompass_Preferred
 View more versions of this product
Product Code. 17158471
  • 433.35€ / 100 tests
Description
  • The OKT4 antibody reacts with CD4, an ∼59 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobulin (Ig) superfamily; CD4 contains four extracellular Ig-like domains (D1 - D4)
  • The epitope for OKT4 has been localized to the D3 domain of the protein, which has a structure resembling an Ig variable domain
  • CD4 is expressed at high levels by most thymocytes and a subpopulation of T cells (T-helper cells), and at lower levels by peripheral blood monocytes and macrophages
  • Moreover, CD4 is a receptor for human immunodeficiency virus (HIV)
  • Binding of the OKT4 antibody to CD4 does not block HIV binding
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Direct Human CD4+ T Cell Isolation Kit (Catalog #19662), EasySep™ Human CD4+ T Cell Enrichment Kit (Catalog #19052), EasySep™ Human CD3 Positive Selection Kit II (Catalog #17851) and EasySep™ Human CD4 Positive Selection Kit II (Catalog #17852)
Specifications
Specifications
CD4
Monoclonal
PerCP-Cyanine5.5
100 Tests
Cynomolgus, Rhesus Monkey, Sooty Mangabey, Chimpanzee, Human
T Cells, T Cells, CD4+, T Cells, Regulatory
Immunohistochemistry, Flow Cytometry, Immunofluorescence, Functional Assay, Immunoprecipitation
OKT4
Mouse
Primary
IgG2b κ