missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse, anti-Human, PerCP-Cyanine5.5, Clone: OKT4, STEMCELL Technologies
Mouse Monoclonal Antibody
193.05€ - 433.35€
Specifications
| Antigen | CD4 |
|---|---|
| Clone | OKT4 |
| Applications | Immunohistochemistry, Flow Cytometry, Immunofluorescence, Functional Assay, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | PerCP-Cyanine5.5 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17168471
|
Stemcell Technologies
60016PS.1 |
25 Tests |
193.05€
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17158471
|
Stemcell Technologies
60016PS |
100 Tests |
433.35€
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The OKT4 antibody reacts with CD4, an ∼59 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobulin (Ig) superfamily; CD4 contains four extracellular Ig-like domains (D1 - D4)
- The epitope for OKT4 has been localized to the D3 domain of the protein, which has a structure resembling an Ig variable domain
- CD4 is expressed at high levels by most thymocytes and a subpopulation of T cells (T-helper cells), and at lower levels by peripheral blood monocytes and macrophages
- Moreover, CD4 is a receptor for human immunodeficiency virus (HIV)
- Binding of the OKT4 antibody to CD4 does not block HIV binding
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Direct Human CD4+ T Cell Isolation Kit (Catalog #19662), EasySep™ Human CD4+ T Cell Enrichment Kit (Catalog #19052), EasySep™ Human CD3 Positive Selection Kit II (Catalog #17851) and EasySep™ Human CD4 Positive Selection Kit II (Catalog #17852)
Specifications
| CD4 | |
| Immunohistochemistry, Flow Cytometry, Immunofluorescence, Functional Assay, Immunoprecipitation | |
| PerCP-Cyanine5.5 | |
| Cynomolgus, Rhesus Monkey, Sooty Mangabey, Chimpanzee, Human | |
| Primary |
| OKT4 | |
| Monoclonal | |
| Mouse | |
| IgG2b κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title