Learn More
CD4 Rat, anti-Mouse, Biotin, Clone: RM4-4, STEMCELL Technologies
Rat Monoclonal Antibody
Brand:  Stemcell Technologies 60029BT
Encompass_Preferred
 View more versions of this product
Product Code. 17148611
  • 517.05€ / 500µg
Description
  • The RM4-4 antibody reacts with mouse CD4, an ∼55 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobin (Ig) superfamily; CD4 contains four extracellular Ig-like domains
  • CD4 is expressed at relatively high levels by most thymocytes and a subpopulation of T cells (T-helper/inducer cells), and at lower levels on dendritic cells
  • In the mouse, CD4 is not expressed by monocytes/macrophages
  • CD4 binds to a non-polymorphic region of MHC II and acts as a co-receptor to the T cell receptor (TCR) in MHC II-restricted antigen recognition by enhancing the avidity of the association between the TCR and MHC II-antigen complex
  • CD4 also functions to amplify signals from the TCR to the cytoplasm through the interaction of its intracellular domain with cytoplasmic tyrosine kinases such as Lck
  • Binding of the RM4-4 antibody does not block binding of antibody clones RM4-5 or GK1.5
Specifications
Specifications
CD4
Monoclonal
Biotin
500 μg
Mouse
T Cells, T Cells, CD4+, T Cells, Regulatory
Flow Cytometry, Functional Assay
RM4-4
Rat
Primary
IgG2b κ