missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Rat, anti-Mouse, Biotin, Clone: RM4-4, STEMCELL Technologies
Rat Monoclonal Antibody
221.40€ - 517.05€
Specifications
| Antigen | CD4 |
|---|---|
| Clone | RM4-4 |
| Applications | Flow Cytometry, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17158611
|
Stemcell Technologies
60029BT.1 |
100 μg |
221.40€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17148611
|
Stemcell Technologies
60029BT |
500 μg |
517.05€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The RM4-4 antibody reacts with mouse CD4, an ∼55 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobin (Ig) superfamily; CD4 contains four extracellular Ig-like domains
- CD4 is expressed at relatively high levels by most thymocytes and a subpopulation of T cells (T-helper/inducer cells), and at lower levels on dendritic cells
- In the mouse, CD4 is not expressed by monocytes/macrophages
- CD4 binds to a non-polymorphic region of MHC II and acts as a co-receptor to the T cell receptor (TCR) in MHC II-restricted antigen recognition by enhancing the avidity of the association between the TCR and MHC II-antigen complex
- CD4 also functions to amplify signals from the TCR to the cytoplasm through the interaction of its intracellular domain with cytoplasmic tyrosine kinases such as Lck
- Binding of the RM4-4 antibody does not block binding of antibody clones RM4-5 or GK1.5
Specifications
| CD4 | |
| Flow Cytometry, Functional Assay | |
| Biotin | |
| Mouse | |
| Primary |
| RM4-4 | |
| Monoclonal | |
| Rat | |
| IgG2b κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title