Learn More
CD45 Mouse, anti-Human, FITC, Clone: 2D1, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60123FI
Description
- The 2D1 antibody reacts with all isoforms of CD45, a type I transmembrane glycoprotein expressed on the surface of hematopoietic stem and progenitor cells and mature blood cells, except mature erythrocytes, platelets, and plasma cells; expression of CD45 is lost during differentiation of these cell lineages
- CD45 is a member of the protein tyrosine phosphatase family and functions in a number of immunoregulatory processes, including cell activation, growth, differentiation, and oncogenic transformation
- The large cytoplasmic portion of CD45 contains two tyrosine phosphatase domains, one of which is enzymatically active, that are involved in modulating the function of intracellular substrates such as the Src kinases Lck and Fyn
- Alternative splicing of three exons (4, 5, and 6) encoding the extracellular RA, RB, and RC polypeptide sequences gives rise to up to 8 isoforms with molecular masses in the range of 180 - 240 kDa
Specifications
| CD45 | |
| Monoclonal | |
| FITC | |
| 100 Tests | |
| Human | |
| Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Leukocytes, Lymphocytes, Myeloid Cells |
| Flow Cytometry | |
| 2D1 | |
| Mouse | |
| Primary | |
| IgG1 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.