missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD48/SLAMF2 Antibody (156-4H9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43403-0.1MG
This item is not returnable.
View return policy
Description
CD48/SLAMF2 Monoclonal specifically detects CD48/SLAMF2 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD48/SLAMF2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry 1:10-1:1000 | |
| BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 156-4H9 | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| CD48 | |
| The immunogen for this antibody was CD48. | |
| 0.1 mg | |
| Cell Biology, Cellular Markers, Immunology, Stem Cell Markers | |
| 962 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction