Learn More
CD55/DAF Antibody (LU30) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-62562
This item is not returnable.
View return policy
Description
CD55/DAF Monoclonal specifically detects CD55/DAF in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.
Specifications
| CD55/DAF | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| CD55 | |
| Human CD55. | |
| 1604 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG λ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| LU30 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, decay accelerating factor for complement, TC | |
| Rabbit | |
| 0.2 mg | |
| The antibody binds to CD55 with a Kd of 13 nM. | |
| Purified |