missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD55/DAF Antibody (LU30) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
261.00€ - 706.00€
Specifications
| Antigen | CD55/DAF |
|---|---|
| Clone | LU30 |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18292914
|
Novus Biologicals
NBP2-62562-0.025MG |
0.025 mg |
261.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692039
|
Novus Biologicals
NBP2-62562 |
0.2 mg |
706.00€
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD55/DAF Monoclonal specifically detects CD55/DAF in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.Specifications
| CD55/DAF | |
| Western Blot 1:100 - 1:2000, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| PBS | |
| 1604 | |
| Human CD55. | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| LU30 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, decay accelerating factor for complement, TC | |
| CD55 | |
| IgG λ | |
| The antibody binds to CD55 with a Kd of 13 nM. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title