missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD69 Armenian Hamster, FITC, Clone: H1.2F3, enQuireBio™
Armenian Hamster Monoclonal Antibody
Brand: EnquireBio QAB90-F-100ug
This item is not returnable.
View return policy
Specifications
| CD69 | |
| Monoclonal | |
| 0.5mg/mL | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| Cd69 | |
| Protein A/G Purified | |
| RUO | |
| 12515 | |
| IgG |
| Flow Cytometry | |
| H1.2F3 | |
| FITC | |
| NP_001028294 | |
| Armenian Hamster | |
| 100 μg | |
| Primary | |
| Store at 2-4°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction