Learn More
Novus Biologicals™ CD7 Antibody (Persongen patent anti-CD7), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28213-1mg
 View more versions of this product
Product Code. 30045225
  • 920.00€ / 1mg
Description
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD7
Monoclonal
Unconjugated
P09564
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Persongen patent anti-CD7
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein
CD7
1 mg
Cytokine Research, Signal Transduction
924