missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CD7 Antibody (Persongen patent anti-CD7), Novus Biologicals™

Human Monoclonal Antibody

213.00€ - 920.00€

Specifications

Antigen CD7
Clone Persongen patent anti-CD7
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30048609
View Documents
Novus Biologicals™
NBP3-28213-50ug
50 μg
213.00€
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30048276
View Documents
Novus Biologicals™
NBP3-28213-100ug
100 μg
338.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30045225
View Documents
Novus Biologicals™
NBP3-28213-1mg
1 mg
920.00€
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications

Specifications

CD7
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Human
Cytokine Research, Signal Transduction
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein
CD7
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Persongen patent anti-CD7
Monoclonal
Purified
RUO
Human
P09564
924
IgG1
Protein A purified
Store at -20°C in powder form. Store at -80°C once reconstituted.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.