missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Antibody (RPA-T8) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NBP2-25195
This item is not returnable.
View return policy
Description
CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, In vitro assay, Flow (Cell Surface), CyTOF-ready.
Specifications
| CD8 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry 0.1-0.25 ug per 1x10^6 cells, Immunohistochemistry, Immunohistochemistry-Frozen reported in scientific literature (PMID 7882470), In vitro assay reported in multiple pieces of scientific literature, Flow (Cell Surface), CyTOF-ready | |
| CD8, CD8 alpha flow, CD8a molecule, LEU2, p32, RPA-T8 RPA-T8 antibody flow, RPA-T8 CD8, RPA-T8 Clone | |
| Mouse | |
| 26 kDa | |
| 0.1 mg | |
| Primary | |
| This product reacts with Human. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), In vitro Assay, CyTOF | |
| RPA-T8 | |
| Unconjugated | |
| NP_741969 | |
| CD8A | |
| CD8 (CD8a; alpha chain) | |
| Protein G purified | |
| RUO | |
| 925 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction