missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Antibody (RPA-T8) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
213.00€ - 480.00€
Specifications
| Antigen | CD8 |
|---|---|
| Clone | RPA-T8 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry 0.1-0.25 ug per 1x10^6 cells, Immunohistochemistry, Immunohistochemistry-Frozen reported in scientific literature (PMID 7882470), In vitro assay reported in multiple pieces of scientific literature, Flow (Cell Surface), CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), In vitro Assay, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18133813
|
Novus Biologicals
NBP2-25195SS |
0.025 mg |
213.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18194262
|
Novus Biologicals
NBP2-25195 |
0.1 mg |
480.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, In vitro assay, Flow (Cell Surface), CyTOF-ready.Specifications
| CD8 | |
| 0.5 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), In vitro Assay, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD8, CD8 alpha flow, CD8a molecule, LEU2, p32, RPA-T8 RPA-T8 antibody flow, RPA-T8 CD8, RPA-T8 Clone | |
| CD8A | |
| IgG1 κ | |
| Protein G purified | |
| This product reacts with Human. |
| RPA-T8 | |
| Flow Cytometry 0.1-0.25 ug per 1x10^6 cells, Immunohistochemistry, Immunohistochemistry-Frozen reported in scientific literature (PMID 7882470), In vitro assay reported in multiple pieces of scientific literature, Flow (Cell Surface), CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| NP_741969 | |
| 925 | |
| CD8 (CD8a; alpha chain) | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 26 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title