missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD81 / TAPA-1 Mouse, Clone: 1.3.3.22, enQuireBio™
Mouse Monoclonal Antibody
199.00€ - 510.00€
Specifications
Antigen | CD81 / TAPA-1 |
---|---|
Clone | 1.3.3.22 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15921308
|
EnquireBio
975-MSM1-P1 |
100 μg |
510.00€
100µg |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15911308
|
EnquireBio
975-MSM1-P0 |
20 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Specifications
CD81 / TAPA-1 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
975 | |
B-Cell line derived from a Burkitt lymphoma | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
26kDa |
1.3.3.22 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_001284578 | |
CD81 | |
IgG1 κ | |
Protein A/G Purified | |
Cell surface | |
Ramos, Daudi, Raji, U266, HUT-78, HUT-102, Jurkat, HL-60, KG1a, A293, A431, MG63, and human lymphocytes. Tonsil or Liver. |