Learn More
CD8a Rat, anti-Mouse, Biotin, Clone: 53-6.7, STEMCELL Technologies
Rat Monoclonal Antibody
Brand:  Stemcell Technologies 60023BT
Encompass_Preferred
 View more versions of this product
Product Code. 17168551
  • 340.20€ / 500µg
Description
  • The 53-6.7 antibody reacts with murine CD8a, a 32 - 34 kDa type I transmembrane glycoprotein which is a subunit of CD8
  • CD8 is a disulfide-bonded dimer, found either as a heterodimer of CD8a (α) and CD8b (β) subunits (i.e., αβ) or a homodimer (αα)
  • CD8 acts as a co-receptor to the T cell receptor (TCR) during T cell activation by binding MHC Class I molecules presented by an antigen-presenting cell
  • It functions to strengthen the association between the TCR and MHC I-antigen complex and to amplify signals from the TCR to the cytoplasm through the interaction of its intracellular domain with cytoplasmic tyrosine kinases such as Lck
  • The CD8a chain binds to the alpha-3 domain of class I MHC molecules
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD8+ T Cell Isolation Kit (Catalog #19853) and EasySep™ Mouse CD90.2 Positive Selection Kit II (Catalog #18951)
Specifications
Specifications
CD8a
Monoclonal
Biotin
500 μg
Mouse, Frog
T Cells, T Cells, CD8+
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay
53-6.7
Rat
Primary
IgG2a κ