missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8a Rat, anti-Mouse, Biotin, Clone: 53-6.7, STEMCELL Technologies
Rat Monoclonal Antibody
109.35€ - 340.20€
Specifications
| Antigen | CD8a |
|---|---|
| Clone | 53-6.7 |
| Applications | CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17178551
|
Stemcell Technologies
60023BT.1 |
50 μg |
109.35€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17168551
|
Stemcell Technologies
60023BT |
500 μg |
340.20€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The 53-6.7 antibody reacts with murine CD8a, a 32 - 34 kDa type I transmembrane glycoprotein which is a subunit of CD8
- CD8 is a disulfide-bonded dimer, found either as a heterodimer of CD8a (α) and CD8b (β) subunits (i.e., αβ) or a homodimer (αα)
- CD8 acts as a co-receptor to the T cell receptor (TCR) during T cell activation by binding MHC Class I molecules presented by an antigen-presenting cell
- It functions to strengthen the association between the TCR and MHC I-antigen complex and to amplify signals from the TCR to the cytoplasm through the interaction of its intracellular domain with cytoplasmic tyrosine kinases such as Lck
- The CD8a chain binds to the alpha-3 domain of class I MHC molecules
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD8+ T Cell Isolation Kit (Catalog #19853) and EasySep™ Mouse CD90.2 Positive Selection Kit II (Catalog #18951)
Specifications
| CD8a | |
| CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
| Biotin | |
| Mouse, Frog | |
| Primary |
| 53-6.7 | |
| Monoclonal | |
| Rat | |
| IgG2a κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title