missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cdc42EP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16697974
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16697974 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16697974 Supplier Invitrogen™ Supplier No. PA5101502

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BORG1.

Cdc42, a small GTPase, regulates Actin polymerization, elongation of cell shape and cell signaling through interactions with many different downstream effector proteins, most of which contain a Cdc42-binding motif known as a CRIB domain. Cdc42EP2 (Cdc42 effector protein 2), also known as BORG1 or CEP2, is a 210 amino acid intracytoplasmic protein that localizes to both the cytoplasm and the cytoskeleton and contains one CRIB domain. Highly expressed in heart and present at lower levels in liver and pancreas, Cdc42EP2 interacts with Cdc42 and is thought to be involved in the organization of the Actin cytoskeleton and may also influence Actin filament assembly and overall cell shape.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc42EP2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CDC42EP2
Gene Accession No. O14613, Q5PQP4, Q8JZX9
Gene Alias 1110008C05Rik; binder of Rho GTPases 1; Borg1; CDC42 effector protein (Rho GTPase binding) 2; CDC42 effector protein 2; CDC42EP2; CEP2; CRIB-containing BOGR1 protein; D19Bwg1013e
Gene Symbols CDC42EP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CDC42EP2(Accession O14613), corresponding to amino acid residues M1-H42.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104252, 10435, 309175
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.