missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDCA4 Polyclonal Antibody
Description
Peptide sequence: MFARGLKRKC VGHEEDVEGA LAGLKTVSSY SLQRQSLLDM SLVKLQLCHM Sequence homology: Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 92%; Rat: 92%; Zebrafish: 100%.
CDCA4 encodes a protein that belongs to the E2F family of transcription factors. This protein regulates E2F-dependent transcriptional activation and cell proliferation, mainly through the E2F/retinoblastoma protein pathway. It also functions in the regulation of JUN oncogene expression. This protein shows distinctive nuclear-mitotic apparatus distribution, it is involved in spindle organization from prometaphase, and may also play a role as a midzone factor involved in chromosome segregation or cytokinesis. Two alternatively spliced transcript variants encoding the same protein have been noted for this gene. Two pseudogenes have also been identified on chromosome 1.
Specifications
Specifications
| Antigen | CDCA4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | CDCA4 |
| Gene Accession No. | Q9BXL8 |
| Gene Alias | 2410018C03Rik; Cdca4; cell division cycle associated 4; cell division cycle-associated protein 4; fb78b08; FLJ20764; Hematopoietic progenitor protein; HEPP; hypothetical protein LOC494037; MGC19517; SEI-3; SEI-3/HEPP; si:dkey-177p2.7; wu:fb78b08; zgc:92217 |
| Gene Symbols | CDCA4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?