missing translation for 'onlineSavingsMsg'
Learn More

CDX2 / Caudal Type Homeobox 2 (GI Epithelial Marker) Mouse, Clone: PCRP-CDX2-1A3, enQuireBio™

Product Code. 15999727
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15999727 100 μg 100µg
15989727 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15999727 Supplier EnquireBio Supplier No. 1045MSM3P1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specifications

Antigen CDX2 / Caudal Type Homeobox 2 (GI Epithelial Marker)
Applications Immunoprecipitation, Western Blot, Immunohistochemistry
Cellular Localization Nuclear
Classification Monoclonal
Clone PCRP-CDX2-1A3
Concentration 200μg/mL
Conjugate Unconjugated
Formulation 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Host Species Mouse
Immunogen Recombinant full-length human CDX2protein
Molecular Weight of Antigen 40kDa
Positive Control HT29 cells or Colon Carcinoma
Purification Method Protein A/G purified from Bioreactor concentrate.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1045
Content And Storage Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.