missing translation for 'onlineSavingsMsg'
Learn More

Cefotiam Dihydrochloride-13C, 15N2, TRC

Product Code. 30356425
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30356425 5 mg 5mg
30349306 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30356425 Supplier Toronto Research Chemicals Supplier No. TRCC2430075MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C1713CH23N715N2O4S3 • 2x( HCl)
Molecular Weight (g/mol) 601.5295
Synonym Cefotiam-13C, 15N2 Hydrochloride, Ceradon-13C, 15N2, HAR-13C, 15N2, HNP-13C, 15N2, Halospor-13C, 15N2, KMS-13C, 15N2, PAS-13C, 15N2, Pansporin-13C, 15N2, Pansporine-13C, 15N2, SEP-13C, 15N2, SFC-13C, 15N2, SFP-13C, 15N2, Spizef-13C, 15N2, Sporidyn-13C, 15N2
IUPAC Name (6R,7R)-7-(2-(2-(amino-15N)thiazol-4-yl-2-13C-3-15N)acetamido)-3-(((1-(2-(dimethylamino)ethyl)-1H-tetrazol-5-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid di hydrogen chloride
SMILES O=C1N([C@@]2([H])[C@@H]1NC(CC3=CS[13C]([15NH2])=[15N]3)=O)C(C(O)=O)=C(CS2)CSC4=NN=NN4CCN(C)C.Cl.Cl

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C18H23N9O4S3.2ClH/c1-25(2)3-4-26-18(22-23-24-26)34-7-9-6-32-15-12(14(29)27(15)13(9)16(30)31)21-11(28)5-10-8-33-17(19)20-10;;/h8,12,15H,3-7H2,1-2H3,(H2,19,20)(H,21,28)(H,30,31);2*1H/t12-,15-;;/m1../s1/i17+1,19+1,20+1;;
Formula Weight 600.0543
Chemical Name or Material Cefotiam Dihydrochloride-13C, 15N2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.