Learn More
CFP Mouse, anti-Human, CoraLite Plus 488, Clone: 1D5A9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66788-100UL
Description
The CFP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CFP. This antibody recognizes human, pig, rat antigen. The CFP antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| CFP | |
| 1D5A9 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Pig, Rat | |
| 155 kDa | |
| IgG1 |
| Western Blot | |
| 1000 μg/mL | |
| BFD, CFP, Complement factor P, complement factor properdin, PFC, PFD, PROPERDIN | |
| 100 μL | |
| 5199 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |