Learn More
CFP Mouse, anti-Human, CoraLite Plus 488, Clone: 1D5A9, Proteintech
Mouse Monoclonal Antibody
145.00€ - 450.00€
Specifications
| Antigen | CFP |
|---|---|
| Clone | 1D5A9 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18098175
|
Proteintech
CL488-66788-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18035295
|
Proteintech
CL488-66788-100UL |
100 μL |
450.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CFP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CFP. This antibody recognizes human, pig, rat antigen. The CFP antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| CFP | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 5199 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1D5A9 | |
| Western Blot | |
| Liquid | |
| Human, Pig, Rat | |
| BFD, CFP, Complement factor P, complement factor properdin, PFC, PFD, PROPERDIN | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.