missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHI3L2 Monoclonal Antibody (632010)

Description
Reconstitute at 0.5 mg/mL in sterile PBS.
CHI3L2 may bind glycan structure with high affinity, but not heparin. Has no chitotriosidase activity.
Specifications
Specifications
| Antigen | CHI3L2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 632010 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with trehalose and no preservative |
| Gene | CHI3L2 |
| Gene Accession No. | Q15782 |
| Gene Alias | CHI3L2; CHIL2; chitinase 3 like 2; chitinase 3-like 2; chitinase-3-like protein 2; Chondrocyte protein 39; YKL39; YKL-39 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?