Learn More
Novus Biologicals™ Chk1 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30982-50ug
 View more versions of this product
Product Code. 30067066
  • 213.00€ / 50µg
Description
Chk1 Polyclonal antibody specifically detects Chk1 in Human,Monkey,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chk1
Polyclonal
Western Blot : 1-10 μg/mL, Immunoprecipitation
Checkpoint, S. pombe, homolog of, 1, CHK1 (checkpoint, S.pombe) homolog, CHK1 checkpoint homolog (S. pombe), CHK1serine/threonine-protein kinase Chk1, EC 2.7.11, EC 2.7.11.1
Sheep were immunized with a protein corresponding to amino acid sequence 1-474 of human Chk1, having a molecular weight of 54 kDa.
50 μg
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Stem Cell Markers, Tumor Suppressors
1111
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Immunoprecipitation, Western Blot
Unconjugated
Phosphate buffered saline
Sheep
Ammonium sulfate precipitation
RUO
Primary
Human, Monkey, Mouse, Rat