missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Chk1 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
213.00€ - 508.00€
Specifications
| Antigen | Chk1 |
|---|---|
| Dilution | Western Blot : 1-10 μg/mL, Immunoprecipitation |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066897
|
Novus Biologicals™
NBP3-30982-250ug |
250 μg |
508.00€
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067066
|
Novus Biologicals™
NBP3-30982-50ug |
50 μg |
213.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chk1 Polyclonal antibody specifically detects Chk1 in Human,Monkey,Mouse,Rat samples. It is validated for Immunoprecipitation,Western BlotSpecifications
| Chk1 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Sheep | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Stem Cell Markers, Tumor Suppressors | |
| Phosphate buffered saline | |
| 1111 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 1-10 μg/mL, Immunoprecipitation | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Monkey, Mouse, Rat | |
| Checkpoint, S. pombe, homolog of, 1, CHK1 (checkpoint, S.pombe) homolog, CHK1 checkpoint homolog (S. pombe), CHK1serine/threonine-protein kinase Chk1, EC 2.7.11, EC 2.7.11.1 | |
| Sheep were immunized with a protein corresponding to amino acid sequence 1-474 of human Chk1, having a molecular weight of 54 kDa. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title