missing translation for 'onlineSavingsMsg'
Learn More

Chlorimuron ethyl, 95%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J66605.03

Quantity 1g

 View more versions of this product

Product Code. 15418029

  • 32.30€ / 1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

90982-32-4
414.817
NSWAMPCUPHPTTC-UHFFFAOYSA-N
56160
ethyl 2-[(4-chloro-6-methoxypyrimidin-2-yl)carbamoylsulfamoyl]benzoate
C15H15ClN4O6S
MFCD00128063
Ethyl 2-(4-chloro-6-methoxy-2-pyrimidinylcarbamoylsulfamoyl)benzoate
CHEBI:47319
CCOC(=O)C1=CC=CC=C1S(=O)(=O)NC(=O)NC2=NC(=CC(=N2)Cl)OC

Specifications

90982-32-4
1g
MFCD00128063
Ethyl 2-(4-chloro-6-methoxy-2-pyrimidinylcarbamoylsulfamoyl)benzoate
NSWAMPCUPHPTTC-UHFFFAOYSA-N
ethyl 2-[(4-chloro-6-methoxypyrimidin-2-yl)carbamoylsulfamoyl]benzoate
56160
414.82
Chlorimuron ethyl
Colorless
C15H15ClN4O6S
14,2093
Freely soluble in aqueous solution
CCOC(=O)C1=CC=CC=C1S(=O)(=O)NC(=O)NC2=NC(=CC(=N2)Cl)OC
414.817
CHEBI:47319
95%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.