missing translation for 'onlineSavingsMsg'
Learn More

Chlorimuron ethyl, 95%, Thermo Scientific™

32.30€ - 88.10€

Chemical Identifiers

CAS 90982-32-4
Molecular Formula C15H15ClN4O6S
Molecular Weight (g/mol) 414.817
MDL Number MFCD00128063
InChI Key NSWAMPCUPHPTTC-UHFFFAOYSA-N
Synonym Ethyl 2-(4-chloro-6-methoxy-2-pyrimidinylcarbamoylsulfamoyl)benzoate
PubChem CID 56160
ChEBI CHEBI:47319
IUPAC Name ethyl 2-[(4-chloro-6-methoxypyrimidin-2-yl)carbamoylsulfamoyl]benzoate
SMILES CCOC(=O)C1=CC=CC=C1S(=O)(=O)NC(=O)NC2=NC(=CC(=N2)Cl)OC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15418029
SDS View Documents
Thermo Scientific Alfa Aesar
J66605.03
1g
32.30€
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15428029
SDS View Documents
Thermo Scientific Alfa Aesar
J66605.06
5g
88.10€
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

90982-32-4
414.817
NSWAMPCUPHPTTC-UHFFFAOYSA-N
56160
ethyl 2-[(4-chloro-6-methoxypyrimidin-2-yl)carbamoylsulfamoyl]benzoate
C15H15ClN4O6S
MFCD00128063
Ethyl 2-(4-chloro-6-methoxy-2-pyrimidinylcarbamoylsulfamoyl)benzoate
CHEBI:47319
CCOC(=O)C1=CC=CC=C1S(=O)(=O)NC(=O)NC2=NC(=CC(=N2)Cl)OC

Specifications

90982-32-4
C15H15ClN4O6S
14,2093
Freely soluble in aqueous solution
CCOC(=O)C1=CC=CC=C1S(=O)(=O)NC(=O)NC2=NC(=CC(=N2)Cl)OC
414.817
CHEBI:47319
95%
Colorless
MFCD00128063
Ethyl 2-(4-chloro-6-methoxy-2-pyrimidinylcarbamoylsulfamoyl)benzoate
NSWAMPCUPHPTTC-UHFFFAOYSA-N
ethyl 2-[(4-chloro-6-methoxypyrimidin-2-yl)carbamoylsulfamoyl]benzoate
56160
414.82
Chlorimuron ethyl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'rtecsNumber' : DG5095000

missing translation for 'tsca' : No

missing translation for 'storageNote' : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.