missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, HPLC Grade, 99.5%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 022922.K2

Quantity 1L

 View more versions of this product

Product Code. 11308197

  • 89.20€ / 1 L
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

108-90-7
112.556
MVPPADPHJFYWMZ-UHFFFAOYSA-N
7964
chlorobenzene
C6H5Cl
MFCD00000530
monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
CHEBI:28097
C1=CC=C(C=C1)Cl

Specifications

108-90-7
1L
Aromatic
C6H5Cl
UN1134
14,2121
Miscible with methanol and diethyl ether. Immiscible with water.
C1=CC=C(C=C1)Cl
112.556
CHEBI:28097
99.5%
Chlorobenzene
1.106 g/mL
28°C (82°F)
1.524
MFCD00000530
605632
monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
MVPPADPHJFYWMZ-UHFFFAOYSA-N
chlorobenzene
7964
112.56
HPLC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.