Learn More
Chlorobenzene, HPLC Grade, 99.5%, Thermo Scientific™
89.20€ - 291.00€
Chemical Identifiers
| CAS | 108-90-7 |
|---|---|
| Molecular Formula | C6H5Cl |
| Molecular Weight (g/mol) | 112.556 |
| MDL Number | MFCD00000530 |
| InChI Key | MVPPADPHJFYWMZ-UHFFFAOYSA-N |
| Synonym | monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene |
| PubChem CID | 7964 |
| ChEBI | CHEBI:28097 |
| IUPAC Name | chlorobenzene |
| SMILES | C1=CC=C(C=C1)Cl |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
11308197
|
Thermo Scientific Alfa Aesar
022922.K2 |
1L |
89.20€
1 L |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
11328197
|
Thermo Scientific Alfa Aesar
022922.M1 |
4 x 1L |
291.00€
4 L |
Estimated Shipment: 08-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
| C6H5Cl | |
| MFCD00000530 | |
| monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene | |
| CHEBI:28097 | |
| C1=CC=C(C=C1)Cl |
Spécification
| 108-90-7 | |
| 28°C (82°F) | |
| 1.524 | |
| MFCD00000530 | |
| 605632 | |
| monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene | |
| MVPPADPHJFYWMZ-UHFFFAOYSA-N | |
| chlorobenzene | |
| 7964 | |
| 112.56 | |
| HPLC |
| 1.106 g/mL | |
| Aromatic | |
| C6H5Cl | |
| UN1134 | |
| 14,2121 | |
| Miscible with methanol and diethyl ether. Immiscible with water. | |
| C1=CC=C(C=C1)Cl | |
| 112.556 | |
| CHEBI:28097 | |
| 99.5% | |
| Chlorobenzene |
Sécurité et traitement
GHS H Statement
H226-H332
Flammable liquid and vapor.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c
H226-H315-H332
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZENE
missing translation for 'einecsNumber' : 203-628-5
missing translation for 'rtecsNumber' : CZ0175000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.