missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl Hexafluoroisopropyl Ether, TRC

Product Code. 30138276
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138276 5 g 5g
30137739 25 g 25g
30006064 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138276 Supplier Toronto Research Chemicals Supplier No. TRCC5878785G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26103-07-1
Molecular Formula C4H3ClF6O
Molecular Weight (g/mol) 216.51
Synonym 2-(Chloromethoxy)-1,1,1,3,3,3-hexafluoropropane, 1,1,1,3,3,3-Hexafluoro-2-chloromethoxypropane, Chloromethyl 1,1,1,3,3,3-Hexafluoro-2-propyl Ether, Chloromethyl-1,1,1,3,3,3-hexafluoroisopropyl Ether
IUPAC Name 2-(chloromethoxy)-1,1,1,3,3,3-hexafluoropropane
SMILES FC(F)(F)C(OCCl)C(F)(F)F

Specifications

Quantity 5 g
InChI Formula InChI=1S/C4H3ClF6O/c5-1-12-2(3(6,7)8)4(9,10)11/h2H,1H2
Formula Weight 215.9777
Chemical Name or Material Chloromethyl Hexafluoroisopropyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.